Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant GACRP-30 (from E. coli)
Human Recombinant GACRP-30 (from E. coli)
Supplier:  VWR International
New Product
Certificates
undefined
Human Recombinant GACRP-30 (from E. coli)
Supplier:  VWR International
New Product

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Endotoxin LAL (EU/µg) ≤1
  • Biological Activity:
    Bioactivity assay: Inhibition of M1 cell proliferation; ED₅₀ ≤2000 ng/ml; Bioactivity ≥500 units/mg
  • Gene ID:
    Q15848
  • Reconstitution Instructions:
    Sterile 10 mM sodium phosphate, 0.5 mM DTT, pH 7.5 at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    Y
  • Protein Synonyms:
    Adiponectin|apm-1|adipolean
  • Protein/Peptide Name:
    GACRP-30 (AF)
  • Purity:
    ≥90%
  • Molecular Weight:
    16.7 kDa
  • Sequence:
    MKGEPGEGAY VYRSAFSVGL ETYVTIPNMP IRFTKIFYNQ QNHYDGSTGK FHCNIPGLYY FAYHITVYMK DVKVSLFKKD KAMLFTYDQY QENNVDQASG SVLLHLEVGD QVWLQVYGEG ERNGLYADND NDSTFTGFLL YHDTN
  • Endotoxin Level:
    Low
  • Formulation:
    10 mM Sodium phosphate, 0.5 mM DTT, pH 7.5
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping Temperature:
    RT
  • Tested Applications:
    Bioassay

Specifications