Order Entry
Puerto Rico
ContactUsLinkComponent
 
Rat Recombinant SHH (from E. coli)
New Product
 
undefined
Rat Recombinant SHH (from E. coli)
New Product

 

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Rat
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Endotoxin LAL (EU/µg) ≤0.1
  • Biological Activity:
    Bioactivity assay: Currently undetermined; ED50 ≤Currently undetermined; Bioactivity ≥NA
  • Gene ID:
    Q63673
  • Reconstitution Instructions:
    Sterile water at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    Y
  • Protein Synonyms:
    Shh|HHG-1
  • Protein/Peptide Name:
    SHH
  • Purity:
    ≥95%
  • Molecular Weight:
    19.9 kDa
  • Sequence:
    MIIGPGRGFG KRQHPKKLTP LAYKQFIPNV AEKTLGASGR YEGKITRNSE RFKELTPNYN PDIIFKDEEN TGADRLMTQR CKDKLNALAI SVMNQWPGVK LRVTEGWDED GHHSEESLHY EGRAVDITTS DRDRSKYGML ARLAVEAGFD WVYYESKARI HCSVKAENSV AAKSDG
  • Endotoxin Level:
    Low
  • Formulation:
    10 mM Sodium Phosphate, pH 7.5
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping Temperature:
    RT