Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Mouse Recombinant FGF-BASIC (from E. coli)

Mouse Recombinant FGF-BASIC (from E. coli)
 
Mouse Recombinant FGF-BASIC (from E. coli)
Mouse Recombinant FGF-BASIC (from E. coli)

 

 

Basic fibroblast growth factor (FGF-basic), also known as FGF-2, is expressed by endothelial cells and is a mediator of angiogenesis. FGF-basic also has cardioprotective functions during heart injury. The application of FGF-basic is a critical component for human embryonic stem cell culture systems and is necessary for maintaining human embryonic stem cells in an undifferentiated state.

  • High quality
  • Low endotoxin
  • Affordable

 : For research use only.

 

 

 

  • Catalog No:
  • 77752-024
  • 77752-026
  • 77752-028
  • 77752-030
  • 77752-032
  • Protein/Peptide Name:
  • FGF-BASIC
  • FGF-BASIC
  • FGF-BASIC
  • FGF-BASIC
  • FGF-BASIC
  • Protein Synonyms:
  • FGF2|HBGF-2|Prostatropin
  • FGF2|HBGF-2|Prostatropin
  • FGF2|HBGF-2|Prostatropin
  • FGF2|HBGF-2|Prostatropin
  • FGF2|HBGF-2|Prostatropin
  • Protein/Peptide Type:
  • Recombinant
  • Recombinant
  • Recombinant
  • Recombinant
  • Recombinant
  • Protein Function:
  • Cytokine
  • Cytokine
  • Cytokine
  • Cytokine
  • Cytokine
  • Species:
  • Mouse
  • Mouse
  • Mouse
  • Mouse
  • Mouse
  • Source:
  • E. coli
  • E. coli
  • E. coli
  • E. coli
  • E. coli
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Gene ID:
  • P15655
  • P15655
  • P15655
  • P15655
  • P15655
  • Sequence:
  • MPALPEDGGA AFPPGHFKDP KRLYCKNGGF FLRIHPDGRV DGVREKSDPH VKLQLQAEER GVVSIKGVCA NRYLAMKEDG RLLASKCVTE ECFFFERLES NNYNTYRSRK YSSWYVALKR TGQYKLGSKT GPGQKAILFL PMSAKS
  • MPALPEDGGA AFPPGHFKDP KRLYCKNGGF FLRIHPDGRV DGVREKSDPH VKLQLQAEER GVVSIKGVCA NRYLAMKEDG RLLASKCVTE ECFFFERLES NNYNTYRSRK YSSWYVALKR TGQYKLGSKT GPGQKAILFL PMSAKS
  • MPALPEDGGA AFPPGHFKDP KRLYCKNGGF FLRIHPDGRV DGVREKSDPH VKLQLQAEER GVVSIKGVCA NRYLAMKEDG RLLASKCVTE ECFFFERLES NNYNTYRSRK YSSWYVALKR TGQYKLGSKT GPGQKAILFL PMSAKS
  • MPALPEDGGA AFPPGHFKDP KRLYCKNGGF FLRIHPDGRV DGVREKSDPH VKLQLQAEER GVVSIKGVCA NRYLAMKEDG RLLASKCVTE ECFFFERLES NNYNTYRSRK YSSWYVALKR TGQYKLGSKT GPGQKAILFL PMSAKS
  • MPALPEDGGA AFPPGHFKDP KRLYCKNGGF FLRIHPDGRV DGVREKSDPH VKLQLQAEER GVVSIKGVCA NRYLAMKEDG RLLASKCVTE ECFFFERLES NNYNTYRSRK YSSWYVALKR TGQYKLGSKT GPGQKAILFL PMSAKS
  • Protease-free:
  • N
  • N
  • N
  • N
  • N
  • Animal-Free:
  • Y
  • Y
  • Y
  • Y
  • Y
  • Nuclease-free:
  • N
  • N
  • N
  • N
  • N
  • Carrier-Free:
  • Y
  • Y
  • Y
  • Y
  • Y
  • Endotoxin-free:
  • N
  • N
  • N
  • N
  • N
  • Endotoxin Level:
  • Low
  • Low
  • Low
  • Low
  • Low
  • Endotoxin Content:
  • Endotoxin LAL (EU/µg) ≤0.1
  • Endotoxin LAL (EU/µg) ≤0.1
  • Endotoxin LAL (EU/µg) ≤0.1
  • Endotoxin LAL (EU/µg) ≤0.1
  • Endotoxin LAL (EU/µg) ≤0.1
  • Biological Activity:
  • Bioactivity assay: 3T3 cell proliferation; ED50 ≤2.5 ng/ml; Bioactivity ≥4.0×10⁵ units/mg
  • Bioactivity assay: 3T3 cell proliferation; ED50 ≤2.5 ng/ml; Bioactivity ≥4.0×10⁵ units/mg
  • Bioactivity assay: 3T3 cell proliferation; ED50 ≤2.5 ng/ml; Bioactivity ≥4.0×10⁵ units/mg
  • Bioactivity assay: 3T3 cell proliferation; ED50 ≤2.5 ng/ml; Bioactivity ≥4.0×10⁵ units/mg
  • Bioactivity assay: 3T3 cell proliferation; ED50 ≤2.5 ng/ml; Bioactivity ≥4.0×10⁵ units/mg
  • Purity:
  • ≥95%
  • ≥95%
  • ≥95%
  • ≥95%
  • ≥95%
  • Grade:
  • RUO
  • RUO
  • RUO
  • RUO
  • RUO
  • Formulation:
  • 10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5
  • 10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5
  • 10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5
  • 10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5
  • 10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5
  • Molecular Weight:
  • 16.5 kDa
  • 16.5 kDa
  • 16.5 kDa
  • 16.5 kDa
  • 16.5 kDa
  • Reconstitution Instructions:
  • Sterile water at 0.1 mg/ml
  • Sterile water at 0.1 mg/ml
  • Sterile water at 0.1 mg/ml
  • Sterile water at 0.1 mg/ml
  • Sterile water at 0.1 mg/ml
  • Storage Conditions:
  • –20 °C
  • –20 °C
  • –20 °C
  • –20 °C
  • –20 °C
  • Shipping Temperature:
  • RT
  • RT
  • RT
  • RT
  • RT
  • Tested Applications:
  • Bioassay
  • Bioassay
  • Bioassay
  • Bioassay
  • Bioassay

 

Product Family Options

 

  • Pack type
    Size
     
     
  • Vial
    100 µg
       
       
    Each
    Each - $321.67
    $321.67
    Each
       $321.67
     

     
     
     
    Specifications
    Cat. No.
    77752-024
    Protein/Peptide Name
    FGF-BASIC
    Protein Synonyms
    FGF2|HBGF-2|Prostatropin
    Protein/Peptide Type
    Recombinant
    Protein Function
    Cytokine
    Species
    Mouse
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    P15655
    Sequence
    MPALPEDGGA AFPPGHFKDP KRLYCKNGGF FLRIHPDGRV DGVREKSDPH VKLQLQAEER GVVSIKGVCA NRYLAMKEDG RLLASKCVTE ECFFFERLES NNYNTYRSRK YSSWYVALKR TGQYKLGSKT GPGQKAILFL PMSAKS
    Protease-free
    N
    Animal-Free
    Y
    Nuclease-free
    N
    Carrier-Free
    Y
    Endotoxin-free
    N
    Endotoxin Level
    Low
    Endotoxin Content
    Endotoxin LAL (EU/µg) ≤0.1
    Biological Activity
    Bioactivity assay: 3T3 cell proliferation; ED50 ≤2.5 ng/ml; Bioactivity ≥4.0×10⁵ units/mg
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5
    Molecular Weight
    16.5 kDa
    Reconstitution Instructions
    Sterile water at 0.1 mg/ml
    Storage Conditions
    –20 °C
    Shipping Temperature
    RT
    Tested Applications
    Bioassay
  • Vial
    10 µg
       
       
    Each
    Each - $70.00
    $70.00
    Each
       $70.00
     

     
     
     
    Specifications
    Cat. No.
    77752-026
    Protein/Peptide Name
    FGF-BASIC
    Protein Synonyms
    FGF2|HBGF-2|Prostatropin
    Protein/Peptide Type
    Recombinant
    Protein Function
    Cytokine
    Species
    Mouse
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    P15655
    Sequence
    MPALPEDGGA AFPPGHFKDP KRLYCKNGGF FLRIHPDGRV DGVREKSDPH VKLQLQAEER GVVSIKGVCA NRYLAMKEDG RLLASKCVTE ECFFFERLES NNYNTYRSRK YSSWYVALKR TGQYKLGSKT GPGQKAILFL PMSAKS
    Protease-free
    N
    Animal-Free
    Y
    Nuclease-free
    N
    Carrier-Free
    Y
    Endotoxin-free
    N
    Endotoxin Level
    Low
    Endotoxin Content
    Endotoxin LAL (EU/µg) ≤0.1
    Biological Activity
    Bioactivity assay: 3T3 cell proliferation; ED50 ≤2.5 ng/ml; Bioactivity ≥4.0×10⁵ units/mg
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5
    Molecular Weight
    16.5 kDa
    Reconstitution Instructions
    Sterile water at 0.1 mg/ml
    Storage Conditions
    –20 °C
    Shipping Temperature
    RT
    Tested Applications
    Bioassay
  • Vial
    1 mg
       
       
    Each
    Each - $1,440.00
    $1,440.00
    Each
       $1,440.00
     

     
     
     
    Specifications
    Cat. No.
    77752-028
    Protein/Peptide Name
    FGF-BASIC
    Protein Synonyms
    FGF2|HBGF-2|Prostatropin
    Protein/Peptide Type
    Recombinant
    Protein Function
    Cytokine
    Species
    Mouse
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    P15655
    Sequence
    MPALPEDGGA AFPPGHFKDP KRLYCKNGGF FLRIHPDGRV DGVREKSDPH VKLQLQAEER GVVSIKGVCA NRYLAMKEDG RLLASKCVTE ECFFFERLES NNYNTYRSRK YSSWYVALKR TGQYKLGSKT GPGQKAILFL PMSAKS
    Protease-free
    N
    Animal-Free
    Y
    Nuclease-free
    N
    Carrier-Free
    Y
    Endotoxin-free
    N
    Endotoxin Level
    Low
    Endotoxin Content
    Endotoxin LAL (EU/µg) ≤0.1
    Biological Activity
    Bioactivity assay: 3T3 cell proliferation; ED50 ≤2.5 ng/ml; Bioactivity ≥4.0×10⁵ units/mg
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5
    Molecular Weight
    16.5 kDa
    Reconstitution Instructions
    Sterile water at 0.1 mg/ml
    Storage Conditions
    –20 °C
    Shipping Temperature
    RT
    Tested Applications
    Bioassay
  • Vial
    500 µg
       
       
    Each
    Each - $828.33
    $828.33
    Each
       $828.33
     

     
     
     
    Specifications
    Cat. No.
    77752-030
    Protein/Peptide Name
    FGF-BASIC
    Protein Synonyms
    FGF2|HBGF-2|Prostatropin
    Protein/Peptide Type
    Recombinant
    Protein Function
    Cytokine
    Species
    Mouse
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    P15655
    Sequence
    MPALPEDGGA AFPPGHFKDP KRLYCKNGGF FLRIHPDGRV DGVREKSDPH VKLQLQAEER GVVSIKGVCA NRYLAMKEDG RLLASKCVTE ECFFFERLES NNYNTYRSRK YSSWYVALKR TGQYKLGSKT GPGQKAILFL PMSAKS
    Protease-free
    N
    Animal-Free
    Y
    Nuclease-free
    N
    Carrier-Free
    Y
    Endotoxin-free
    N
    Endotoxin Level
    Low
    Endotoxin Content
    Endotoxin LAL (EU/µg) ≤0.1
    Biological Activity
    Bioactivity assay: 3T3 cell proliferation; ED50 ≤2.5 ng/ml; Bioactivity ≥4.0×10⁵ units/mg
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5
    Molecular Weight
    16.5 kDa
    Reconstitution Instructions
    Sterile water at 0.1 mg/ml
    Storage Conditions
    –20 °C
    Shipping Temperature
    RT
    Tested Applications
    Bioassay
  • Vial
    50 µg
       
       
    Each
    Each - $178.33
    $178.33
    Each
       $178.33
     

     
     
     
    Specifications
    Cat. No.
    77752-032
    Protein/Peptide Name
    FGF-BASIC
    Protein Synonyms
    FGF2|HBGF-2|Prostatropin
    Protein/Peptide Type
    Recombinant
    Protein Function
    Cytokine
    Species
    Mouse
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    P15655
    Sequence
    MPALPEDGGA AFPPGHFKDP KRLYCKNGGF FLRIHPDGRV DGVREKSDPH VKLQLQAEER GVVSIKGVCA NRYLAMKEDG RLLASKCVTE ECFFFERLES NNYNTYRSRK YSSWYVALKR TGQYKLGSKT GPGQKAILFL PMSAKS
    Protease-free
    N
    Animal-Free
    Y
    Nuclease-free
    N
    Carrier-Free
    Y
    Endotoxin-free
    N
    Endotoxin Level
    Low
    Endotoxin Content
    Endotoxin LAL (EU/µg) ≤0.1
    Biological Activity
    Bioactivity assay: 3T3 cell proliferation; ED50 ≤2.5 ng/ml; Bioactivity ≥4.0×10⁵ units/mg
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5
    Molecular Weight
    16.5 kDa
    Reconstitution Instructions
    Sterile water at 0.1 mg/ml
    Storage Conditions
    –20 °C
    Shipping Temperature
    RT
    Tested Applications
    Bioassay