Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant Myostatin (from E. coli)
Human Recombinant Myostatin (from E. coli)
Supplier:  VWR International
New Product
Certificates
undefined
Human Recombinant Myostatin (from E. coli)
Supplier:  VWR International
New Product

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage Conditions:
    −20 °C
  • Endotoxin Content:
    Endotoxin LAL (EU/µg) ≤0.1
  • Biological Activity:
    Bioactivity assay: MPC-11 cell cytotoxicity; ED₅₀ ≤50 ng/ml; Bioactivity ≥2.0×10⁴ units/mg
  • Gene ID:
    O14793
  • Reconstitution Instructions:
    Sterile 20 mM HCl at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    Y
  • Protein Synonyms:
    GDF-8|MSTN
  • Protein/Peptide Name:
    Myostatin
  • Purity:
    ≥95%
  • Molecular Weight:
    12.4/24.8 kDa
  • Sequence:
    #N/A
  • Endotoxin Level:
    Low
  • Formulation:
    0.1% Trifluoroacetic acid (TFA)
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping Temperature:
    RT
  • Tested Applications:
    Bioassay

Specifications