Human Recombinant FGF-10 (from E. coli)
Supplier: VWR International
New Product
Certificates
About this item
Fibroblast growth factor 10 (FGF-10) is a growth factor that is important during embryonic development, especially during lung, limb, brain, heart, and kidney morphogenesis. FGF-10 is expressed in mesenchymal cells and facilitates epithelial-mesenchymal signaling through binding the epithelially expressed FGF receptor 2b (FGFR2b). FGF-10 also functions as a mitogen for keratinizing epidermal cells, and induces the migration and invasion of cancer cells.
High quality
- Low endotoxin
- Affordable
Caution:
For research use only.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Storage Conditions:−20 °C
- Endotoxin Content:Endotoxin LAL (EU/µg) ≤0.1
- Biological Activity:Bioactivity assay: 4MBr-5 cell proliferation; ED₅₀ ≤200 ng/ml; Bioactivity ≥5.0×10³ units/mg
- Gene ID:O15520
- Reconstitution Instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-Free:Y
- Protease-free:N
- Animal-Free:N
- Protein Synonyms:Keratinocyte growth factor 2
- Protein/Peptide Name:FGF-10
- Purity:≥95%
- Molecular Weight:19.3 kDa
- Sequence:MLGQDMVSPE ATNSSSSSFS SPSSAGRHVR SYNHLQGDVR WRKLFSFTKY FLKIEKNGKV SGTKKENCPY SILEITSVEIGVVAVKAINS NYYLAMNKKG KLYGSKEFNN DCKLKERIEE NGYNTYASFN WQHNGRQMYV ALNGKGAPRR GQKTRRKNTS AHFLPMVVHS
- Endotoxin Level:Low
- Formulation:10 mM Sodium phosphate, pH 7.5
- Nuclease-free:N
- Grade:RUO
- Shipping Temperature:RT
- Tested Applications:Bioassay