Human Recombinant TARC (from E. coli)
Supplier: VWR International
New Product
Certificates
About this item
Thymus and activation regulated chemokine (TARC), also known as CCL17, is a chemokine that is constitutively produced by thymus tissue and activated peripheral blood mononuclear cells (PBMCs), including dendritic cells. TARC signals through the CCR4 receptor to induce chemotaxis of Type 2 T helper (Th2) cells. TARC is important in asthma and allergic diseases, along with bacterial and viral infections.
- High quality
- Low endotoxin
- Affordable
Caution:
For research use only.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Endotoxin LAL (EU/µg) ≤0.1
- Biological Activity:Bioactivity assay: Currently undetermined; ED50 ≤Currently undetermined; Bioactivity ≥5.0×10³ units/mg
- Gene ID:Q92583
- Reconstitution Instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-Free:Y
- Protease-free:N
- Animal-Free:N
- Protein Synonyms:CCL17|thymus and activation regulated chemokine|ABCD-2|SCYA17
- Protein/Peptide Name:TARC
- Purity:≥95%
- Molecular Weight:8.1
- Sequence:ARGTNVGREC CLEYFKGAIP LRKLKTWYQT SEDCSRDAIV FVTVQGRAIC SDPNNKRVKN AVKYLQSLER S
- Endotoxin Level:Low
- Formulation:0.1% Trifluoroacetic Acid (TFA)
- Nuclease-free:N
- Grade:RUO
- Shipping Temperature:RT