Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant Rank Ligand (from E. coli)
Human Recombinant Rank Ligand (from E. coli)
Supplier:  VWR International
New Product
Certificates
undefined
Human Recombinant Rank Ligand (from E. coli)
Supplier:  VWR International
New Product

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Endotoxin LAL (EU/µg) ≤1
  • Biological Activity:
    Bioactivity assay: Ability to induce bioluminescence in engineered RANKL bioassay cells ; ED50 ≤50 ng/ml; Bioactivity ≥2.0×10⁴ units/mg
  • Gene ID:
    O14788
  • Reconstitution Instructions:
    Sterile water at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    Y
  • Protein Synonyms:
    TNFSF11|TRANCE|OPGL|ODF
  • Protein/Peptide Name:
    Rank ligand
  • Purity:
    ≥95%
  • Molecular Weight:
    19.7
  • Sequence:
    EKAMVDGSWL DLAKRSKLEA QPFAHLTINA TDIPSGSHKV SLSSWYHDRG WAKISNMTFS NGKLIVNQDG FYYLYANICF RHHETSGDLA TEYLQLMVYV TKTSIKIPSS HTLMKGGSTK YWSGNSEFHF YSINVGGFFK LRSGEEISIE VSNPSLLDPD QDATYFGAFK VRDID
  • Endotoxin Level:
    Low
  • Formulation:
    10 mM Sodium phosphate, pH 7.5
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping Temperature:
    RT

Specifications