Human Recombinant Rank Ligand (from E. coli)
Supplier: VWR International
New Product
Certificates
About this item
Receptor activator of nuclear factor kappa-B Ligand (RANK Ligand) is a cell-bound marker related to the tumor necrosis factor (TNF) family of proteins. RANK Ligand plays a critical role in bone metabolism and osteoclast differentiation. T cell expression of RANK Ligand promotes dendritic cell maturation.
- High quality
- Low endotoxin
- Affordable
Caution:
For research use only.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Endotoxin LAL (EU/µg) ≤1
- Biological Activity:Bioactivity assay: Ability to induce bioluminescence in engineered RANKL bioassay cells ; ED50 ≤50 ng/ml; Bioactivity ≥2.0×10⁴ units/mg
- Gene ID:O14788
- Reconstitution Instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-Free:Y
- Protease-free:N
- Animal-Free:N
- Protein Synonyms:TNFSF11|TRANCE|OPGL|ODF
- Protein/Peptide Name:Rank ligand
- Purity:≥95%
- Molecular Weight:19.7
- Sequence:EKAMVDGSWL DLAKRSKLEA QPFAHLTINA TDIPSGSHKV SLSSWYHDRG WAKISNMTFS NGKLIVNQDG FYYLYANICF RHHETSGDLA TEYLQLMVYV TKTSIKIPSS HTLMKGGSTK YWSGNSEFHF YSINVGGFFK LRSGEEISIE VSNPSLLDPD QDATYFGAFK VRDID
- Endotoxin Level:Low
- Formulation:10 mM Sodium phosphate, pH 7.5
- Nuclease-free:N
- Grade:RUO
- Shipping Temperature:RT