Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant TH9 Cell Polarizing PB (from E. coli cells)
Human Recombinant TH9 Cell Polarizing PB (from E. coli cells)
Catalog # 77753-620
Supplier:  VWR International
New Product
CAS Number:  
undefined
Human Recombinant TH9 Cell Polarizing PB (from E. coli cells)
Catalog # 77753-620
Supplier:  VWR International
New Product
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    Each
  • Storage Conditions:
    At or below –20 °C
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    N
  • Protein/Peptide Name:
    HUMAN TH9 CELL POLARIZING PB
  • Endotoxin Level:
    Low
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping Temperature:
    RT

Specifications

About this item

Cytokines for polarizing human T cells into Th9 cells. Th9 cells are a unique subset of T helper cells that produce IL-9. An important function of IL-9 producing T cells is fighting inflammation and helminth infections. Th9 cells can be generated from naive CD4+ T cells in culture by culturing them in the presence of IL-4 and TGF-β 1. It also appears that IFNγ and IL-27 can inhibit the generation of Th9 cells.

  • High quality
  • Low endotoxin
  • Affordable
Caution: For research use only.