Order Entry
Puerto Rico
ContactUsLinkComponent
Macaque Recombinant EBI3 SUBUNIT (IL-27/IL-35) (from E. coli cells)
Macaque Recombinant EBI3 SUBUNIT (IL-27/IL-35) (from E. coli cells)
Supplier:  VWR International
New Product
Certificates
undefined
Macaque Recombinant EBI3 SUBUNIT (IL-27/IL-35) (from E. coli cells)
Supplier:  VWR International
New Product

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Macaque
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Endotoxin LAL (EU/µg) ≤0.1
  • Biological Activity:
    Bioactivity: No biological activity data is available at this time
  • Gene ID:
    102144621 (gene ID)
  • Reconstitution Instructions:
    Sterile 20 mM HCl at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    N
  • Protein Synonyms:
    IL-35/EBI3|IL-27 EBI3 subunit|IL-35 EBI3 subunit
  • Protein/Peptide Name:
    EBI3 SUBUNIT (IL-27/IL-35)
  • Purity:
    ≥95%
  • Molecular Weight:
    23.4 kDa
  • Sequence:
    MRKGPPAALT LPRVQCRAPR YPIAVDCSWT LPPAPNSTSP VSFIATYRFG MAARGHSWPC LQQTPASTSC TIADVRLFSM APYVLNVTAV HPWGSSSSFV PFIAEHIIKP DPPEGVRLSP LAERQLQVQW EPPRSWPFPE IFSLKYWIRY KRQGAARFHQ VGPIEATSFI LRAVRPRARY CVQVAAQDLT DYGELSDWSL PATTPMSPGK
  • Endotoxin Level:
    Low
  • Formulation:
    0.1% Trifluoroacetic Acid (TFA)
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping Temperature:
    RT

Specifications