Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant PDGF-BB (from E. coli)
Human Recombinant PDGF-BB (from E. coli)
Supplier:  VWR International
New Product
Certificates
undefined
Human Recombinant PDGF-BB (from E. coli)
Supplier:  VWR International
New Product

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage Conditions:
    At or below –20 °C
  • Endotoxin Content:
    Endotoxin LAL (EU/µg) ≤0.01
  • Biological Activity:
    Bioactivity assay: Proliferation of NR6R-3T3 ; ED50 ≤5 ng/ml; Bioactivity ≥1.6 x10³ IU/µg
  • Gene ID:
    P01127
  • Reconstitution Instructions:
    Sterile water at 0.1-0.5 mg/ml
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    Y
  • Protein Synonyms:
    GDGF|ODGF
  • Protein/Peptide Name:
    PDGF-BB
  • Purity:
    ≥98%
  • Molecular Weight:
    12.4/24.9 kDa
  • Sequence:
    MSLGSLTIAE PAMIAECKTR TEVFEISRRL IDRTNANFLV WPPCVEVQRC SGCCNNRNVQ CRPTQVQLRP VQVRKIEIVR KKPIFKKATV TLEDHLACKC ETVAAARPVT
  • Endotoxin Level:
    Low
  • Formulation:
    10 mM sodium phosphate, pH 7.5
  • Nuclease-free:
    N
  • Grade:
    GMP
  • Shipping Temperature:
    Ambient
  • Tested Applications:
    Bioassay

Specifications