Human Recombinant OSM (from E. coli)
Supplier: VWR International
New Product
Certificates
About this item
Oncostatin M is a cytokine that is produced by macrophages, dendritic cells, and T lymphocytes during inflammatory events. The Type-I and Type-II Oncostatin M receptors are located on the cell surface of endothelial and tumor cells, contain the glycoprotein 130 (gp130) subunit, and activate the JAK/STAT signaling pathway.
- High quality
- Low endotoxin
- Affordable
Oncostatin M functions to inhibit tumor cell proliferation, induce liver stem cell maturation, regulate cytokine production during hematopoiesis and inflammation, stimulate bone formation, and promote nervous system development.
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Endotoxin LAL (EU/µg) ≤1
- Biological Activity:Bioactivity assay: TF-1 cell proliferation; ED50 ≤4 ng/ml; Bioactivity ≥2.5×10⁵ units/mg
- Gene ID:P13725
- Reconstitution Instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-Free:Y
- Protease-free:N
- Animal-Free:N
- Protein Synonyms:Onc-M
- Protein/Peptide Name:OSM
- Purity:≥95%
- Molecular Weight:23.8 kDa
- Sequence:MAAIGSCSKE YRVLLGQLQK QTDLMQDTSR LLDPYIRIQG LDVPKLREHC RERPGAFPSE ETLRGLGRRG FLQTLNATLGCVLHRLADLE QRLPKAQDLE RSGLNIEDLE KLQMARPNIL GLRNNIYCMA QLLDNSDTAE PTKAGRGASQ PPTPTPASDA FQRKLEGCRFLHGYHRFMHS VGRVFSKWGE SPNRSRRHSP HQALRKGVRR
- Endotoxin Level:Low
- Formulation:10 mM sodium phosphate, pH 7.5
- Nuclease-free:N
- Grade:RUO
- Shipping Temperature:RT
- Tested Applications:Bioassay