Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse Anti-Nipah Virus Fusion Glycoprotein Antibody IgG Titer Serologic Assay Kit
Mouse Anti-Nipah Virus Fusion Glycoprotein Antibody IgG Titer Serologic Assay Kit
Catalog # 77761-664
Supplier:  ReVacc Scientific
New Product
CAS Number:  
undefined
Mouse Anti-Nipah Virus Fusion Glycoprotein Antibody IgG Titer Serologic Assay Kit
Catalog # 77761-664
Supplier:  ReVacc Scientific
Supplier Number:  E60G23
New Product
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Indirect
  • Conjugate ELISA:
    Unconjugated
  • Host:
    Goat
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    Nipah virus fusion glycoprotein
  • Size:
    96 Tests

Specifications

About this item

This kit is developed for serologic test for mouse IgG titer of Anti-NiV F&G antibody in serum/plasma or purified mouse antibody (monoclonal and polyclonal) in vitro. It is for research use only.

  • Hendra virus (HeV) and Nipah virus (NiV) are henipaviruses (HNVs), which are members of the paramyxovirus family, including a number of pathogens causing disease in humans such as measles, mumps and parainfluenza viruses. After a primary infection, HeV and NiV enter the target cells with a receptor-binding glycoprotein (G) and a fusion glycoprotein (F). F and G are the main targets of vaccine development, associated with neutralizing antibody response and correlated of protection. A rapid and effective assay kit detecting the levels of anti-NiV F and G in mouse serum can facilitate research on antibodies produced in response to vaccine candidates or viral challenge.

Assay Principles: 
This kit is developed for a standard indirect-ELISA format, providing a rapid detection of anti-NiV fusion glycoprotein (F&G) mouse IgG in serum. The kit consists of high-bind detachable 96-well plate pre-coated with F&G protein, Positive control, Negative Control, an HRP-Anti-Mouse IgG secondary antibody, TMB and dilution/wash/stop buffer.

Use protocol (4 simple steps):

1. Detach wells from plate based on your samples and keep at room temperature. Dilute your samples, positive and negative controls in sample dilution buffer and add them to wells (0.1 ml/well). Incubate at 37 °C for 1 hour or room temperature for 1.5 hour. Wash wells 4 times by wash solution (0.3 ml/well).
2. Dilute the Secondary antibody HRP-Anti-Mouse IgG by wash solution and add to each well (0.1 ml/well). Incubate at room temperature for 0.5 - 1 hour. Avoid light by foil. Wash wells as mentioned.
3. Add TMB to each well (0.1 ml/well) and incubate at room temperature for 10 - 30 minutes. 
4. Stop the substrate reaction by adding stop solution (0.1 ml/well).  Absorbance (OD) is calculated as the absorbance at 450 nm. The OD Value reflects the amount of antibody bound.

Caution:

For research use only.