Order Entry
Puerto Rico
ContactUsLinkComponent
 
Human Recombinant IL-34 (from CHO cells)
New Product
 
undefined
Human Recombinant IL-34 (from CHO cells)
New Product

 

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/Peptide Type:
    Recombinant
  • Source:
    CHO cells
  • Species:
    Human
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Endotoxin LAL (EU/µg) ≤0.1
  • Biological Activity:
    Bioactivity assay: Currently undetermined; ED50 ≤Currently undetermined; Bioactivity ≥NA
  • Gene ID:
    NP_689669
  • Reconstitution Instructions:
    Sterile water at 0.1 mg/ml (reconstitution at concentrations >0.1 mg/ml may result in a precipitate)
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    N
  • Protein/Peptide Name:
    IL-34
  • Purity:
    ≥95%
  • Molecular Weight:
    26 kDa
  • Sequence:
    NEPLEMWPLT QNEECTVTGF LRDKLQYRSR LQYMKHYFPI NYKISVPYEG VFRIANVTRL QRAQVSEREL RYLWVLVSLS ATESVQDVLL EGHPSWKYLQ EVETLLLNVQ QGLTDVEVSP KVESVLSLLN APGPNLKLVR PKALLDNCFR VMELLYCSCC KQSSVLNWQD CEVPSPQSCS PEPSLQYAAT QLYPPPPWSP SSPPHSTGSV RPVRAQGEGL LPHHHHHH
  • Endotoxin Level:
    Low
  • Formulation:
    Phosphate buffered saline (PBS)
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping Temperature:
    RT