Order Entry
Puerto Rico
ContactUsLinkComponent
 
Mouse Recombinant IL-10 (from E. coli)
New Product
 
undefined
Mouse Recombinant IL-10 (from E. coli)
New Product

 

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Endotoxin LAL (EU/µg) ≤1
  • Biological Activity:
    Bioactivity assay: MC/9 cell proliferation; ED50 ≤5 ng/ml; Bioactivity ≥2.0×10⁵ units/mg
  • Gene ID:
    P18893
  • Reconstitution Instructions:
    Sterile water at 0.1 mg/ml (reconstitution at concentrations >0.1 mg/ml may result in a precipitate)
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    N
  • Protein Synonyms:
    Cytokine synthesis inhibitory factor|CSIF
  • Protein/Peptide Name:
    IL-10
  • Purity:
    ≥95%
  • Molecular Weight:
    18.9 kDa
  • Sequence:
    MSRGQYSRED NNCTHFPVGQ SHMLLELRTA FSQVKTFFQT KDQLDNILLT DSLMQDFKGY LGCQALSEMI QFYLVEVMPQ AEKHGPEIKE HLNSLGEKLK TLRMRLRRCH RFLPCENKSK AVEQVKSDFN KLQDQGVYKA MNEFDIFINC IEAYMMIKMK S
  • Endotoxin Level:
    Low
  • Formulation:
    10 mM sodium phosphate, pH 7.5
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping Temperature:
    RT
  • Tested Applications:
    Bioassay