Mouse Recombinant EXODUS-2/CCL21 (from E. coli)
New Product
Exodus-2, also known as CCL21 and 6Ckine, is a chemokine that is strongly produced in the human lymph nodes and spleen. Exodus-2 signals through the chemokine receptor CCR7 to regulate thymocyte and activated T cell migration. Exodus-2 also mediates the homing of lymphocytes to the lymphatic system. Human and mouse exodus-2 proteins share greater than 85% amino acid sequence identity.
High quality
- Low endotoxin
- Affordable
: For research use only.
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Mouse
- Storage Conditions:–20 °C
- Endotoxin Content:Endotoxin LAL (EU/µg) ≤0.1
- Biological Activity:Bioactivity: No biological activity data is available at this time
- Gene ID:P84444
- Reconstitution Instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-Free:Y
- Protease-free:N
- Animal-Free:N
- Protein Synonyms:CCL21|6Ckine
- Protein/Peptide Name:EXODUS-2/CCL21
- Purity:≥95%
- Molecular Weight:12 kDa
- Sequence:SDGGGQDCCL KYSQKKIPYS IVRGYRKQEP SLGCPIPAIL FSPRKHSKPE LCANPEEGWV QNLMRRLDQP PAPGKQSPGC RKNRGTSKSG KKGKGSKGCK RTEQTQPSRG
- Endotoxin Level:Low
- Formulation:0.1% Trifluoroacetic acid (TFA)
- Nuclease-free:N
- Grade:RUO
- Shipping Temperature:RT