Order Entry
Puerto Rico
ContactUsLinkComponent
 
Human Recombinant IGF-I (from E. coli)
New Product
 
undefined
Human Recombinant IGF-I (from E. coli)
New Product

 

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Endotoxin LAL (EU/µg) ≤0.1
  • Biological Activity:
    Bioactivity assay: Human breast cancer cell line MCF-7; ED50 ≤1.5 ng/ml; Bioactivity ≥6.7×10⁵ units/mg
  • Gene ID:
    P05019
  • Reconstitution Instructions:
    Sterile water at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    N
  • Protein Synonyms:
    Somatomedin C|mechano growth factor|IGF-IA
  • Protein/Peptide Name:
    IGF-I
  • Purity:
    ≥95%
  • Molecular Weight:
    7.7 kDa
  • Sequence:
    GPETLCGAEL VDALQFVCGD RGFYFNKPTG YGSSSRRAPQ TGIVDECCFR SCDLRRLEMY CAPLKPAKSA
  • Endotoxin Level:
    Low
  • Formulation:
    0.1% Trifluoroacetic acid (TFA)
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping Temperature:
    RT
  • Tested Applications:
    Bioassay