Autoclavable Tyvek Breather Bags Tubing, Continuous Form
New Product
A significant upgrade over paper-based steam sterilization tubing. Cut tubing to desired length and heat seal both ends closed.
- Manufactured and packaged in an ISO certified cleanroom Peelable seals Recommended for use in conjunction with a secondary Tyvek™ sterilization pouch Autoclavable at 121 to 125 °C Tear and puncture resistant Hydrophobic Lint-free Heat seals closed Tubing provides sizing flexibility
The medical Grade 1073B Tyvek® material has the highest microbial barrier properties of any steam sterilization compatible materials. Tyvek tubing is also compatible with VHP Decontamination cycles commonly used with Isolator technology. Will not absorb or adsorb water.
Closure type: Heat seal
Opening: Peel
:
Supplied with Certificate of Conformance.