Human Recombinant TH17 CELL POLARIZING PB (from E. coli)
Catalog # 77753-612
Supplier: VWR International
New Product
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Size:Each
- Storage Conditions:At or below –20 °C
- Endotoxin-free:N
- Carrier-Free:Y
- Protease-free:N
- Animal-Free:N
- Protein/Peptide Name:TH17 CELL POLARIZING PB
- Sequence:0
- Endotoxin Level:Low
- Nuclease-free:N
- Grade:RUO
- Shipping Temperature:RT
Specifications
About this item
Cytokines for polarizing human T cells into Th17 cells. T helper 17 cells (Th17) are a distinct type of T cell subset that are thought to be important in immunity against microbes at the epitilial and mucosal interfaces of the body. Disruption of the regular function or balance of Th17 cells is thought to be involved in several autoimmune diseases and/or opportunistic infections. Th17 cells are characterized by their ability to produce IL-17, IL-22 and IL-21. Th17 cells can be cultured from naive CD4+ T cells by culturing them in the presence of anti-CD3 and anti-CD28 stimulation with recombinant IL-6 and recombinant TGF-β 1.
High quality
Low endotoxin
Affordable
Caution: For research use only.