Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant IL-3 (from E. coli)
Human Recombinant IL-3 (from E. coli)
Supplier:  VWR International
New Product
Certificates
undefined
Human Recombinant IL-3 (from E. coli)
Supplier:  VWR International
New Product

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage Conditions:
    At or below –20 °C
  • Endotoxin Content:
    Endotoxin LAL (EU/µg) ≤0.05
  • Biological Activity:
    Bioactivity assay: Proliferation of TF-1; ED50 ≤2 ng/ml; Bioactivity ≥4.2 x10³ IU/µg
  • Gene ID:
    P08700
  • Reconstitution Instructions:
    Sterile water at 0.1 - 0.5 mg/ml
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    Y
  • Protein Synonyms:
    MCGF|Multi-CSF|HCGF|P-cell stimulating factor
  • Protein/Peptide Name:
    IL-3
  • Purity:
    ≥97%
  • Molecular Weight:
    15.2 kDa
  • Sequence:
    MAPMTQTTPL KTSWVNCSNM IDEIITHLKQ PPLPLLDFNN LNGEDQDILM ENNLRRPNLE AFNRAVKSLQ NASAIESILK NLLPCLPLAT AAPTRHPIHI KDGDWNEFRR KLTFYLKTLE NAQAQQTTLS LAIF
  • Endotoxin Level:
    Low
  • Formulation:
    10 mM sodium phosphate, 150 mM sodium chloride, pH 7.5
  • Nuclease-free:
    N
  • Grade:
    GMP
  • Shipping Temperature:
    Ambient
  • Tested Applications:
    Bioassay

Specifications