Mouse Recombinant IL-11 (from E. coli)
Supplier: VWR International
New Product
Certificates
About this item
Interleukin 11 (IL-11) is a member of the gp130 family of cytokines. IL-11 functions to promote hematopoietic stem cell proliferation and megakaryocyte differentiation. In non-hematopoietic cell populations, IL-11 stimulates acute-phase proteins, modulates the development of immunoglobulin-producing B cells, and regulates bone turnover. IL-11 binds the IL-11Rα receptor to activate JAK downstream signaling.
High quality
Low endotoxin
Affordable
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Mouse
- Storage Conditions:–20 °C
- Endotoxin Content:Endotoxin LAL (EU/µg) ≤1
- Biological Activity:Bioactivity assay: B9 cell proliferation; ED50 ≤250 ng/ml; Bioactivity ≥4.0×10³units/mg
- Gene ID:P47873
- Reconstitution Instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-Free:Y
- Protease-free:N
- Animal-Free:Y
- Protein Synonyms:AGIF (Adipogenesis Inhibitory Factor)|oprelvekin
- Protein/Peptide Name:IL-11
- Purity:≥95%
- Molecular Weight:19.3 kDa
- Sequence:MPGPPAGSPR VSSDPRADLD SAVLLTRSLL ADTRQLAAQM RDKFPADGDH SLDSLPTLAM SAGTLGSLQL PGVLTRLRVD LMSYLRHVQW LRRAGGPSLK TLEPELGALQ ARLERLLRRL QLLMSRLALP QAAPDQPVIP LGPPASAWGS IRAAHAILGG LHLTLDWAVR GLLLLKTRL
- Endotoxin Level:Low
- Formulation:0.1% Trifluoroacetic Acid (TFA)
- Nuclease-free:N
- Grade:RUO
- Shipping Temperature:RT
- Tested Applications:Bioassay