Mouse Recombinant IL-16 (from E. coli)
Supplier: VWR International
New Product
Certificates
About this item
Interleukin 16 (IL-16) is produced by CD4+ and CD8+ T cells and functions as a chemoattractant for lymphocytes, monocytes, eosinophils, dendritic cells, and langerhans cells. Additionally, IL-16 upregulates the expression of the interleukin 2 receptor (CD25), modulates T lymphocyte growth, and suppresses the replication of the human immunodeficiency virus (HIV) and simian immunodeficiency viruses (SIV).
High quality
Low endotoxin
Affordable
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Mouse
- Storage Conditions:–20 °C
- Endotoxin Content:Endotoxin LAL (EU/µg) ≤0.1
- Biological Activity:Bioactivity: No biological activity data is available at this time
- Gene ID:O54824
- Reconstitution Instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-Free:Y
- Protease-free:N
- Animal-Free:N
- Protein Synonyms:Lymphocyte Chemoattractant Factor (LCF)
- Protein/Peptide Name:IL-16
- Purity:≥95%
- Molecular Weight:13.2 kDa
- Sequence:MHDLNSSTDS AASASAASDI SVESKEATVC TVTLEKTSAG LGFSLEGGKG SLHGDKPLTI NRIFKGDRTG EMVQPGDEIL QLAGTAVQGL TRFEAWNVIK ALPDGPVTIV IRRTSLQCKQ TTASADS
- Endotoxin Level:Low
- Formulation:10 mM sodium phosphate, pH 7.5
- Nuclease-free:N
- Grade:RUO
- Shipping Temperature:RT