- Pack type:Vial
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Size:Each
- Storage Conditions:At or below –20 °C
- Endotoxin-free:N
- Carrier-Free:Y
- Protease-free:N
- Animal-Free:N
- Protein/Peptide Name:MONOCYTE/DC PB
- Endotoxin Level:Low
- Nuclease-free:N
- Grade:RUO
- Shipping Temperature:RT
Cytokines for the differentation of human monycyte-derived dendritic cells. Dendritic cells are an important type of immune cell that function to activate T and B cells via their role as an antigen presenting cell. Monocytes isolated from peripheral blood can be differentiated to immature dendritic cells (iDCs) through culturing in the presence of interleukin-4 (IL-4) and granulocyte-macrophage colony stimulating factor (GM-CSF). The characteristics of monocyte-derived iDCs are detectable after culture with IL-4/GM-CSF and include: the down regulation of CD14 surface expression, an increase in dextran uptake and an increased response to MIP-1α. Subsequent culturing with pro-inflammatory cytokines (such as TNF-α, IL-1β, and IL-6) can further differentiate the iDCs into mature dendritic cells (mDCs) that have full T cell stimulating capacity. This bundle includes 1X 100 μg of Recombinant Human IL-4 and 1X 100 μg of Recombinant Human GM-CSF.
- High quality Low endotoxin Affordable