Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant FOLLISTATIN (from E. coli)
Human Recombinant FOLLISTATIN (from E. coli)
Supplier:  VWR International
New Product
Certificates
undefined
Human Recombinant FOLLISTATIN (from E. coli)
Supplier:  VWR International
New Product

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Endotoxin LAL (EU/µg) ≤0.1
  • Biological Activity:
    Bioactivity assay: Neutralization of human activin A induced cytotoxicity of MPC-11 cells; ED50 ≤200 ng/ml; Bioactivity ≥5.0×10³ units/mg
  • Gene ID:
    P19883
  • Reconstitution Instructions:
    Sterile water at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    N
  • Protein Synonyms:
    FS|activin-binding protein|FSH-suppressing protein (FSP)
  • Protein/Peptide Name:
    FOLLISTATIN
  • Purity:
    ≥95%
  • Molecular Weight:
    31.7 kDa
  • Sequence:
    MGNCWLRQAK NGRCQVLYKT ELSKEECCST GRLSTSWTEE DVNDNTLFKW MIFNGGAPNC IPCKETCENV DCGPGKKCRMNKKNKPRCVC APDCSNITWK GPVCGLDGKT YRNECALLKA RCKEQPELEV QYQGRCKKTC RDVFCPGSST CVVDQTNNAY CVTCNRICPEPASSEQYLCG NDGVTYSSAC HLRKATCLLG RSIGLAYEGK CIKAKSCEDI QCTGGKKCLW DFKVGRGRCS LCDELCPDSK SDEPVCASDNATYASECAMK EAACSSGVLL EVKHSGSCN
  • Endotoxin Level:
    Low
  • Formulation:
    10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping Temperature:
    RT
  • Tested Applications:
    Bioassay

Specifications