Human Recombinant HB-EGF (from E. coli)
New Product
Heparin-binding EGF-like growth factor (HB-EGF) is a member of the epidermal growth factor (EGF) family and is expressed by monocytes and macrophages. HB-EGF is the predominant growth factor involved in epithelialization during wound healing. HB-EGF signals through the receptor tyrosine kinase ErbB2 to maintain adult heart homeostasis, and promotes cardiac valve development through binding in high affinity to the epidermal growth factor receptor (EGFR). HB-EGF binds the the ErbB4 receptor tyrosine kinase to mediate implantation of the human blastocyst. HB-EGF also functions as a potent mitogen for fibroblasts and smooth muscle cells.
High quality
Low endotoxin
Affordable
: For research use only.
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Endotoxin LAL (EU/µg) ≤0.1
- Biological Activity:Bioactivity assay: 3T3 cell proliferation; ED50 ≤1 ng/ml; Bioactivity ≥1.0×10⁶ units/mg
- Gene ID:Q99075
- Reconstitution Instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-Free:Y
- Protease-free:N
- Animal-Free:N
- Protein Synonyms:HBEGF|Diphtheria toxin receptor|DT-R
- Protein/Peptide Name:HB-EGF
- Purity:≥95%
- Molecular Weight:9.9 kDa
- Sequence:MDLQEADLDL LRVTLSSKPQ ALATPNKEEH GKRKKKGKGL GKKRDPCLRK YKDFCIHGEC KYVKELRAPS CICHPGYHGERCHGLSL
- Endotoxin Level:Low
- Formulation:10 mM sodium phosphate, pH 7.5
- Nuclease-free:N
- Grade:RUO
- Shipping Temperature:RT
- Tested Applications:Bioassay