Order Entry
Puerto Rico
ContactUsLinkComponent
 
Rat Recombinant GDNF (from E. coli)
New Product
 
undefined
Rat Recombinant GDNF (from E. coli)
New Product

 

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Rat
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Endotoxin LAL (EU/µg) ≤1
  • Biological Activity:
    Bioactivity: No biological activity data is available at this time
  • Gene ID:
    Q07731
  • Reconstitution Instructions:
    Sterile water at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    N
  • Protein Synonyms:
    ATF-1
  • Protein/Peptide Name:
    GDNF
  • Purity:
    ≥95%
  • Molecular Weight:
    15.1/30.1 kDa
  • Sequence:
    MSPDKQAAAL PRRERNRQAA AASPENSRGK GRRGQRGKNR GCVLTAIHLN VTDLGLGYET KEELIFRYCS GSCEAAETMY DKILKNLSRS RRLTSDKVGQ ACCRPVAFDD DLSFLDDSLV YHILRKHSAK RCGCI
  • Endotoxin Level:
    Low
  • Formulation:
    10 mM sodium phosphate, pH 7.5
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping Temperature:
    RT