Human Recombinant FGF-21 (from E. coli)
Supplier: VWR International
New Product
Certificates
About this item
Fibroblast growth factor 21 (FGF-21) is an endocrine hormone that regulates energy homeostasis and exerts cardioprotective functions during heart injury. FGF-21 is expressed in the liver, pancreas, heart, and adipose tissues. FGF-21 signaling is activated through the FGFR1c receptor and β-Klotho co-receptor. FGF-21 is an important regulator of glucose uptake and reduces cell apoptosis under stress conditions.
High quality
Low endotoxin
Affordable
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Endotoxin LAL (EU/µg) ≤0.1
- Biological Activity:Bioactivity assay: NIH-3T3 proliferation with β-klotho and heparin; ED50 ≤500 ng/ml; Bioactivity ≥2.0×10³ unit/mg
- Gene ID:Q9NSA1
- Reconstitution Instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-Free:Y
- Protease-free:N
- Animal-Free:Y
- Protein Synonyms:FGFL
- Protein/Peptide Name:FGF-21
- Purity:≥95%
- Molecular Weight:19.5 kDa
- Sequence:MHPIPDSSPL LQFGGQVRQR YLYTDDAQQT EAHLEIREDG TVGGAADQSP ESLLQLKALK PGVIQILGVK TSRFLCQRPD GALYGSLHFD PEACSFRELL LEDGYNVYQS EAHGLPLHLP GNKSPHRDPA PRGPARFLPL PGLPPALPEP PGILAPQPPD VGSSDPLSMV GPSQGRSPSY AS
- Endotoxin Level:Low
- Formulation:10 mM sodium phosphate, 100 mM sodium chloride, pH 7.5
- Nuclease-free:N
- Grade:RUO
- Shipping Temperature:RT
- Tested Applications:Bioassay