Order Entry
Puerto Rico
ContactUsLinkComponent
 
Human, Mouse, Rat Recombinant GDF-11 (from E. coli)
New Product
 
undefined
Human, Mouse, Rat Recombinant GDF-11 (from E. coli)
New Product

 

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
    Mouse
    Rat
  • Storage Conditions:
    At or below –20 °C
  • Endotoxin Content:
    Endotoxin LAL (EU/µg) ≤0.1
  • Biological Activity:
    Bioactivity assay: Alkaline phosphatase activity in ATDC5 cells; ED50 ≤100 ng/ml; Bioactivity ≥1.0×10⁴ units/mg
  • Gene ID:
    O95390/Q9Z1W4/Q9Z217
  • Reconstitution Instructions:
    Sterile 10 mM HCl at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    N
  • Protein Synonyms:
    BMP-11
  • Protein/Peptide Name:
    GDF-11
  • Purity:
    ≥95%
  • Molecular Weight:
    12.5/24.9 kDa
  • Sequence:
    NLGLDCDEHS SESRCCRYPL TVDFEAFGWD WIIAPKRYKA NYCSGQCEYM FMQKYPHTHL VQQANPRGSA GPCCTPTKMS PINMLYFNDK QQIIYGKIPG MVVDRCGCS
  • Endotoxin Level:
    Low
  • Formulation:
    0.1% trifluoroacetic acid (TFA)
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping Temperature:
    RT
  • Tested Applications:
    Bioassay