Mouse Recombinant IL-2 (from E. coli)
Supplier: VWR International
New Product
Certificates
About this item
Interleukin 2 (IL-2) is an immunomodulatory cytokine that is produced by lymphocytes. IL-2 signals through the IL-2R receptor to induce activated T cell proliferation and promote T cell differentiation. IL-2 also stimulates the proliferation and differentiation of B cells, natural killer cells, monocytes, and macrophages.
- High quality
- Low endotoxin
- Affordable
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Mouse
- Storage Conditions:–20 °C
- Endotoxin Content:Endotoxin LAL (EU/µg) ≤0.1
- Biological Activity:Bioactivity assay: CTLL-2 cell proliferation; ED50 ≤5 ng/ml; Bioactivity ≥2.0×10⁵ units/mg
- Gene ID:P04351
- Reconstitution Instructions:Sterile 10 mM acetic acid at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-Free:Y
- Protease-free:N
- Animal-Free:N
- Protein Synonyms:T cell growth factor|TCGF|Aldesleukin
- Protein/Peptide Name:IL-2
- Purity:≥95%
- Molecular Weight:17.4 kDa
- Sequence:MAPTSSSTSS STAEAQQQQQ QQQQQQQHLE QLLMDLQELL SRMENYRNLK LPRMLTFKFY LPKQATELKD LQCLEDELGP LRHVLDLTQS KSFQLEDAEN FISNIRVTVV KLKGSDNTFE CQFDDESATV VDFLRRWIAF CQSIISTSPQ
- Endotoxin Level:Low
- Formulation:10 mM sodium citrate, pH 4.0
- Nuclease-free:N
- Grade:RUO
- Shipping Temperature:RT
- Tested Applications:Bioassay