Human Recombinant G-CSF (from E. coli)
Supplier: VWR International
New Product
Certificates
About this item
Granulocyte-colony stimulating factor (G-CSF) is a cytokine that functions as a potent inducer of neutrophilic granulocyte proliferation, terminal differentiation, and activation. G-CSF synthesis occurs in monocyte, macrophage, epithelial, endothelial, and fibroblast cells after activation by bacterial endotoxins, tumor necrosis factor alpha (TNF-α), interleukin 1 (IL-1), or interleukin 17 (IL-17).
- High quality
- Low endotoxin
- Affordable
The functional activity of G-CSF is mediated through the granulocyte colony-stimulating factor receptor (G-CSF-R) to activate JAK/STAT and MAPK signal transduction pathways. G-CSF also promotes neurogenesis and inhibits neuronal apoptosis. Human and mouse G-CSF proteins are cross-reactive.
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Endotoxin LAL (EU/µg) ≤0.1
- Biological Activity:Bioactivity assay: NFS-60 cell proliferation; ED50 ≤0.060 ng/ml; Bioactivity ≥1.7×10⁷ units/mg
- Gene ID:P09919
- Reconstitution Instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-Free:Y
- Protease-free:N
- Animal-Free:Y
- Protein Synonyms:CSF-3|MGI-1G|GM-CSFβ|pluripoietin
- Protein/Peptide Name:G-CSF
- Purity:≥95%
- Molecular Weight:18.8 kDa
- Sequence:MTPLGPASSL PQSFLLKCLE QVRKIQGDGA ALQEKLCATY KLCHPEELVL LGHSLGIPWA PLSSCPSQAL QLAGCLSQLH SGLFLYQGLL QALEGISPEL GPTLDTLQLD VADFATTIWQ QMEELGMAPA LQPTQGAMPA FASAFQRRAG GVLVASHLQS FLEVSYRVLR HLAQP
- Endotoxin Level:Low
- Formulation:20 mM acetic acid, 50 mM sodium chloride
- Nuclease-free:N
- Grade:RUO
- Shipping Temperature:RT
- Tested Applications:Bioassay