Order Entry
Puerto Rico
ContactUsLinkComponent
 
Human Recombinant TGF-B 1 (from CHO cells)
New Product
 
undefined
Human Recombinant TGF-B 1 (from CHO cells)
New Product

 

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/Peptide Type:
    Recombinant
  • Source:
    CHO cells
  • Species:
    Human
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Endotoxin LAL (EU/µg) ≤0.1
  • Biological Activity:
    Bioactivity assay: Inhibition of IL-4-induced HT-2 cell proliferation; ED50 ≤0.5 ng/ml; Bioactivity ≥2.0×10⁶ units/mg
  • Gene ID:
    P01137
  • Reconstitution Instructions:
    Sterile 10 mM HCl at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    N
  • Protein Synonyms:
    Differentiation inhibiting factor|cartilage-inducing factor
  • Protein/Peptide Name:
    TGF-B 1
  • Purity:
    ≥95%
  • Molecular Weight:
    12.8/25.6 kDa
  • Sequence:
    ALDTNYCFSS TEKNCCVRQL YIDFRKDLGW KWIHEPKGYH ANFCLGPCPY IWSLDTQYSK VLALYNQHNP GASAAPCCVP QALEPLPIVY YVGRKPKVEQ LSNMIVRSCK CS
  • Endotoxin Level:
    Low
  • Formulation:
    0.1% trifluoroacetic acid (TFA) and Trehalose in a 20:1 Trehalose to protein ratio
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping Temperature:
    RT
  • Tested Applications:
    Bioassay