Human Recombinant IL-31 (from E. coli)
New Product
Interleukin 31 (IL-31) is an immunoregulatory cytokine that is expressed by activated type 2 T helper (Th2) cells. IL-31 signals through a heterodimer receptor consisting of the IL-31 Receptor A (IL-31RA) and the oncostatin M receptor (OSMR), which are expressed on monocytes, epithelial cells, and keratinocytes. IL-31 promotes allergic reactions and inflammatory skin diseases.
- High quality
- Low endotoxin
- Affordable
: For research use only.
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Endotoxin LAL (EU/µg) ≤0.1
- Biological Activity:Bioactivity assay: STAT3 Activation in U-87 MG cells; ED50 ≤Detectable at ≤25 ng/ml; Bioactivity ≥0
- Gene ID:Q6EBC2
- Reconstitution Instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-Free:Y
- Protease-free:N
- Animal-Free:Y
- Protein/Peptide Name:IL-31
- Purity:≥95%
- Molecular Weight:15.8 kDa
- Sequence:SHTLPVRLLR PSDDVQKIVE ELQSLSKMLL KDVEEEKGVL VSQNYTLPCL SPDAQPPNNI HSPAIRAYLK TIRQLDNKSV IDEIIEHLDK LIFQDAPETN ISVPTDTHEC KRFILTISQQ FSECMDLALK SLTSGAQQAT T
- Endotoxin Level:Low
- Formulation:20 mM sodium phosphate, 50 mM sodium chloride, pH 7.5
- Nuclease-free:N
- Grade:RUO
- Shipping Temperature:RT
- Tested Applications:Bioassay