Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant IL-17AF (from E. coli)
Human Recombinant IL-17AF (from E. coli)
Supplier:  VWR International
New Product
Certificates
undefined
Human Recombinant IL-17AF (from E. coli)
Supplier:  VWR International
New Product

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Endotoxin LAL (EU/µg) ≤1
  • Biological Activity:
    Bioactivity assay: Production of IL-6 from mouse 3T3 cells; ED50 ≤50 ng/ml; Bioactivity ≥2.0×10⁴ units/mg
  • Gene ID:
    Q16552/Q96PD4
  • Reconstitution Instructions:
    Sterile water at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    N
  • Protein/Peptide Name:
    IL-17AF
  • Purity:
    ≥95%
  • Molecular Weight:
    30.7 kDa
  • Sequence:
    IL-17A:MIVKAGITIP RNPGCPNSED KNFPRTVMVN LNIHNRNTNT NPKRSSDYYN RSTSPWNLHR NEDPERYPSV IWEAKCRHLG CINADGNVDY HMNSVPIQQE ILVLRREPPH CPNSFRLEKI LVSVGCTCVT PIVHHVA IL-17F:MRKIPKVGHT FFQKPESCPP VPGGSMKLDI GIINENQRVS MSRNIESRST SPWNYTVTWD PNRYPSEVVQ AQCRNLGCIN AQGKEDISMN SVPIQQETLV VRRKHQGCSV SFQLEKVLVT VGCTCVTPVI HHVQ
  • Endotoxin Level:
    Low
  • Formulation:
    0.1% trifluoroacetic acid (TFA)
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping Temperature:
    RT
  • Tested Applications:
    Bioassay

Specifications