Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant IL-2 (from E. coli)
Human Recombinant IL-2 (from E. coli)
Supplier:  VWR International
New Product
Certificates
undefined
Human Recombinant IL-2 (from E. coli)
Supplier:  VWR International
New Product

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Endotoxin LAL (EU/µg) ≤0.1
  • Biological Activity:
    Bioactivity assay:CTLL-2 cell proliferation; ED50 ≤3 ng/ml; Bioactivity ≥3.3×10⁵ units/mg
  • Gene ID:
    P60568
  • Reconstitution Instructions:
    Sterile 10 mM HCl at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    N
  • Protein Synonyms:
    T cell growth factor|TCGF|Aldesleukin
  • Protein/Peptide Name:
    IL-2
  • Purity:
    ≥97%
  • Molecular Weight:
    15.5 kDa
  • Sequence:
    MAPTSSSTKK TQLQLEHLLL DLQMILNGIN NYKNPKLTRM LTFKFYMPKK ATELKHLQCL EEELKPLEEV LNLAQSKNFH LRPRDLISNI NVIVLELKGS ETTFMCEYAD ETATIVEFLN RWITFCQSII STLT
  • Endotoxin Level:
    Low
  • Formulation:
    0.1% Trifluoroacetic acid (TFA)
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping Temperature:
    RT
  • Tested Applications:
    Bioassay

Specifications