About this item
This kit is developed for serologic test for human IgA titer of Anti-RSV F antibody in serum/plasma or purified human antibody (monoclonal and polyclonal) in vitro. Antibodies of IgM and IgA class are a sign of an active infection (primary infection and reactivation) and disappear during convalescence. In some cases they can persist for several months.
These kits were tested to bind to monoclonal antibodies in IgA format (M96T49) or human sera
- Recommendations - customers who intend to use this kit to generate quantitative curves are suggested to consider the monoclonal antibodies to generate the reference curve: Recombinant Human Anti-RSV Fusion IgA Antibody (M96T49)
Respiratory syncytial virus (RSV) annually causes approximately three million acute lower respiratory infections in children and is typified by recurrent infections in infants. Two major RSV subtypes, A and B, circulate with 1 to 2 epidemic periods per year. The RSV fusion (F) glycoprotein is a dominant antigen and a prominent vaccine target. A rapid and effective assay kit detecting the levels of anti-RSV F from both subtypes A and B in human serum can facilitate research on antibodies produced in response to RSV infection or vaccine candidates.
Assay principles:
This kit is developed for a standard indirect-ELISA format, providing a rapid detection of anti-RSV fusion (F) glycoprotein human IgA in serum. The kit consists of high-bind detachable 96-well plate pre-coated with trimeric F protein, positive control, negative control, an HRP-Anti-Human IgA secondary antibody, TMB and dilution/wash/stop buffer.
Use protocol (Four simple steps):
1. Detach wells from plate based on your samples and keep at room temperature. Dilute your samples, positive and negative controls in sample dilution buffer and add them to wells (0.1 ml/well). Incubate at 37 °C for 1 hour or room temperature for 1.5 hour. Wash wells four times by wash solution (0.3 ml/well).
2. Dilute the Secondary antibody HRP-Anti-Human IgA by wash solution and add to each well (0.1 ml/well). Incubate at room temperature for 0.5 to 1 hour. Avoid light by foil. Wash wells as mentioned.
3. Add TMB to each well (0.1 ml/well) and incubate at room temperature for 10 to 30 minutes.
4. Stop the substrate reaction by adding stop solution (0.1 ml/well). Absorbance (OD) is calculated as the absorbance at 450 nm. The OD Value reflects the amount of antibody bound.
For research use only.
Specifications
- Assay duration:Multiple steps
- Assay Type (ELISA with LOV):Indirect
- Conjugate ELISA:Unconjugated
- Format:Pre-coated
- Host:Goat
- Primary antibody reactivity:Human
- Target protein:Respiratory Syncytial Virus (RSV) Fusion (F) glycoprotein from both subtype A and B