Order Entry
Puerto Rico
ContactUsLinkComponent
Human Anti-SARS-CoV-2 Spike (Trimer) IgA Titer Serologic Assay Kit
Human Anti-SARS-CoV-2 Spike (Trimer) IgA Titer Serologic Assay Kit
Catalog # 77761-654
Supplier:  ReVacc Scientific
New Product
CAS Number:  
undefined
Human Anti-SARS-CoV-2 Spike (Trimer) IgA Titer Serologic Assay Kit
Catalog # 77761-654
Supplier:  ReVacc Scientific
Supplier Number:  D43L97
New Product
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Indirect
  • Conjugate ELISA:
    Unconjugated
  • Format:
    Pre-coated
  • Host:
    Goat
  • Primary antibody reactivity:
    Human
  • Target protein:
    Severe Acute Respiratory Syndrome Coronavirus 2 (SARS-CoV-2) trimeric spike
  • Size:
    96 tests

Specifications

About this item

This kit is developed for serologic test for human IgA titer of Anti-SARS-CoV-2 trimeric Spike (Wuhan Strain) antibody in serum/plasma or purified human antibody (monoclonal and polyclonal) in vitro. Antibodies of IgM and IgA class are a sign of an active infection (primary infection and reactivation) and disappear during convalescence. In some cases they can persist for several months.

  • These kits were tested to bind to monoclonal antibodies in IgA format (Q74F36) or human sera

  • Recommendations - customers who intend to use this kit to generate quantitative curves are suggested to consider the monoclonal antibodies to generate the reference curve: Recombinant Human Anti-SARS or SARS-CoV-2 S RBD IgA Antibody (Q74F36)

Severe Acute Respiratory Syndrome Coronavirus 2 (SARS-CoV-2) caused the pandemic of COVID-19. A rapid and effective assay kit detecting the levels of anti-SARS-CoV-2 Spike in human serum can facilitate research on antibodies produced in response to SARS-CoV-2 infection or vaccines.

Assay principles:
This kit is developed for a standard indirect-ELISA format, providing a rapid detection of anti- Severe Acute Respiratory Syndrome Coronavirus 2 (SARS-CoV-2) trimeric Spike (Wuhan Strain) human IgA in serum. The kit consists of high-bind detachable 96-well plate pre-coated with trimeric Spike protein, positive control, negative control, an HRP-Anti-Human IgA secondary antibody, TMB and dilution/wash/stop buffer.

Use protocol (Four simple steps):
1. Detach wells from plate based on your samples and keep at room temperature. Dilute your samples, positive and negative controls in sample dilution buffer and add them to wells (0.1 ml/well). Incubate at 37 °C for 1 hour or room temperature for 1.5 hour. Wash wells four times by wash solution (0.3 ml/well).
2. Dilute the secondary antibody HRP-Anti-Human IgA by wash solution and add to each well (0.1 ml/well). Incubate at room temperature for 0.5 to 1 hour. Avoid light by foil. Wash wells as mentioned.
3. Add TMB to each well (0.1 ml/well) and incubate at room temperature for 10 to 30 minutes.
4. Stop the substrate reaction by adding stop solution (0.1 ml/well). Absorbance (OD) is calculated as the absorbance at 450 nm. The OD value reflects the amount of antibody bound.

Caution:

For research use only.