ContactUsLinkComponent
Brady® SPC® Absorbents Base Neutralizing Polymer
Brady® SPC® Absorbents Base Neutralizing Polymer
Catalog # 77775-246
Supplier:  Brady Worldwide
New Product
Brady® SPC® Absorbents Base Neutralizing Polymer
Catalog # 77775-246
Supplier:  Brady Worldwide
Supplier Number:  123065
New Product
Specifications
  • Description:
    Base neutralizing polymer
Specifications
About this item
The base neutralizer is used to solidify a wide range of base spills.
  • Class: chemical
  • Container type: bottle
Base neutralizer used for most bases. Polymers neutralize and change color to let users know neutralization has taken place. These two pound shakers are non-toxic, non-corrosive, and non-hazardous. Base neutralizer turns orange to indicate neutralization is complete. Minimum order quantitiy: 10.