ContactUsLinkComponent
Extended Length LfTS fit Pipette Tip Reload Systems, Non Sterile
Extended Length LfTS fit Pipette Tip Reload Systems, Non Sterile
Catalog # 77873-988
Supplier:  VistaLab
New Product
undefined
Extended Length LfTS fit Pipette Tip Reload Systems, Non Sterile
Catalog # 77873-988
Supplier:  VistaLab
Supplier Number:  4080-1042
New Product
Specifications
  • Volume:
    1 - 10 µl
  • Length:
    50.04 mm
  • Tip color:
    Clear
  • Packed:
    960 tips/reload
  • Sterile:
    -
  • Tip style:
    Extended length
  • For use with:
    LfTS, LTS
Specifications
About this item
Designed to provide a low-force tip solution for more comfortable pipetting; less force needed to acquire tips.
  • LfTS reload systems offer a convenient and economical way to refill empty tip racks, saving both time and money
  • Reload systems dramatically reduce plastic waste by reusing empty racks
  • Fine tip for improved accuracy and precision
  • Polypropylene
These 10 μl extended length tips have a maximum volume of 20 μl and are compatible with 2, 10, and 20 μl pipettes. LfTS reload system tips, wafers, and racks are fully autoclavable.