Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-SARS and SARS-CoV-2 Human Recombinant Monoclonal Antibody [clone: RV88]
Anti-SARS and SARS-CoV-2 Human Recombinant Monoclonal Antibody [clone: RV88]
Supplier:  ReVacc Scientific
New Product
Certificates
undefined
Anti-SARS and SARS-CoV-2 Human Recombinant Monoclonal Antibody [clone: RV88]
Supplier:  ReVacc Scientific
New Product

Specifications

  • Catalog No:
  • 77761-720
  • 77761-718
  • Antigen Symbol:
  • SARS and SARS-CoV-2
  • SARS and SARS-CoV-2
  • Antigen Name:
  • Severe Acute Respiratory Syndrome Coronavirus (SARS, 2003 strain) and Severe Acute Respiratory Syndrome Coronavirus 2 (SARS-CoV-2)
  • Severe Acute Respiratory Syndrome Coronavirus (SARS, 2003 strain) and Severe Acute Respiratory Syndrome Coronavirus 2 (SARS-CoV-2)
  • Antigen Synonyms:
  • Spike receptor-binding domain (RBD)
  • Spike receptor-binding domain (RBD)
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Recombinant Monoclonal
  • Recombinant Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • RV88
  • RV88
  • Reactivity:
  • Human
  • Human
  • Host:
  • Human
  • Human
  • Isotype:
  • IgA1 kappa
  • IgA1 kappa
  • Western Blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Flow Cytometry:
  • Yes
  • Yes
  • Neutralisation:
  • Yes
  • Yes
  • Concentration:
  • 1 mg/ml by refractometry
  • 1 mg/ml by refractometry
  • Storage Buffer:
  • PBS, pH 7.2
  • PBS, pH 7.2
  • Storage Temperature:
  • Store at 4 °C, −20 °C
  • Store at 4 °C, −20 °C
  • Shipping Temperature:
  • Ice
  • Ice

Specifications

Frequently Bought Together