This antibody has been tested to bind to His tagged protein.
Type: Primary
Antigen: His tag
Clonality: Recombinant monoclonal
Clone: RV7
Conjugation:
Epitope: 6-His (HHHHHH) or 8-His or 10-His tag
Host: Sheep
Isotype: IgG1 kappa
Reactivity: Sheep
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- 77761-670
- 77761-672
- Antigen Symbol:
- His tag
- His tag
- Antigen Name:
- Polyhistidine-tag
- Polyhistidine-tag
- Antibody Type:
- Primary
- Primary
- Clonality:
- Recombinant Monoclonal
- Recombinant Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- RV7
- RV7
- Reactivity:
- Sheep
- Sheep
- Host:
- Sheep
- Sheep
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Flow Cytometry:
- Yes
- Yes
- Neutralisation:
- Yes
- Yes
- Epitope:
- 6-His (HHHHHH) or 8-His or 10-His tag
- 6-His (HHHHHH) or 8-His or 10-His tag
- Purification:
- Purified by Protein C chromatography
- Purified by Protein C chromatography
- Concentration:
- 1 mg/ml by refractometry
- 1 mg/ml by refractometry
- Storage Buffer:
- PBS, pH 7.2
- PBS, pH 7.2
- Storage Temperature:
- Store at 4 °C, −20 °C
- Store at 4 °C, −20 °C
- Shipping Temperature:
- Ice
- Ice