About this item
This antibody has been tested to bind to Varicella-Zoster virus (VZV) gE protein (Cat. No. 77394-666). An example of comparison of IgM and IgG using size exclusion chromatography is shown.
Type: Primary
Antigen: VZV
Clonality: Recombinant Monoclonal
Clone: RV37
Conjugation: Unconjugated
Epitope:
Host: Human
Isotype: IgM kappa
Reactivity: Human
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77761-750
- 77761-748
- Antigen Symbol:
- VZV
- VZV
- Antigen Name:
- Varicella-Zoster virus
- Varicella-Zoster virus
- Antigen Synonyms:
- Glycoprotein E (gE) protein
- Glycoprotein E (gE) protein
- Antibody Type:
- Primary
- Primary
- Clonality:
- Recombinant Monoclonal
- Recombinant Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- RV37
- RV37
- Reactivity:
- Human
- Human
- Host:
- Human
- Human
- Isotype:
- IgM kappa
- IgM kappa
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Flow Cytometry:
- Yes
- Yes
- Neutralisation:
- Yes
- Yes
- Concentration:
- 1 mg/ml by Refractometry
- 1 mg/ml by Refractometry
- Storage Buffer:
- PBS, pH 7.2
- PBS, pH 7.2
- Storage Temperature:
- Store at 4 °C, −20 °C
- Store at 4 °C, −20 °C
- Shipping Temperature:
- Ice
- Ice