Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Proteometer-L Buffer Packets

Proteometer-L Buffer Packets

Proteometer-L Buffer Packets (7X)
Catalog # 77775-512
Supplier:   NOVILYTIC, LLC
Supplier #:  BUF-PL-001
Proteometer-L Buffer Packets
Catalog # 77775-512
Supplier:  NOVILYTIC, LLC
CAS Number:  
Proteometer-L Buffer Packets
Proteometer-L Buffer Packets

Proteometer-L Buffer Packets (7X)

CAS Number:  
Price
$512.00
Log in for availability
Pack of 1
Product Details & Documents

7 sealed buffer packets specifically formulated for use with Proteometer-L kit.

  • Buffer packets available for reorder when the Proteometer-L reactor and/or Proteometer-L reagent are not required

Intended for use with the Proteometer-L kit.

Ordering information: Reorder individually as needed.

Packaging: Supplied in a custom box with instructions for use. Store at room temperature.

Specifications

Specifications

  • Description:
    Proteometer-L Buffer Packets (7X)

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.