Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Proteometer-L Consumables Kit

Proteometer-L Consumables Kit

Proteometer-L consumables kit, reagent to be stored at -20 °C
Catalog # 77775-506
Supplier:   NOVILYTIC, LLC
Supplier #:  KIT-PL-005-01
Proteometer-L Consumables Kit
Catalog # 77775-506
Supplier:  NOVILYTIC, LLC
CAS Number:  
Proteometer-L Consumables Kit
Proteometer-L Consumables Kit

Proteometer-L consumables kit, reagent to be stored at -20 °C

CAS Number:  
Price
$2,304.00
Log in for availability
Kit of 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

The Proteometer-L Kit transforms any LC into an analytical system capable of identifying and quantifying titer and relative aggregate content in crude culture filtrate in <10 minutes, without sample preparation.

  • mAb titer + aggregate measurement: One assay for two critical attributes

  • No Protein A purification required: Reduces costs
  • No sample pretreatment: Reduces hours of sample prep
  • Direct analysis from CFB: HCP removal is not needed
  • No method development: Reduces time until method development
  • Plug-and-play kit for any LC: Easy adoption - no software needed
  • Low sample volume (≤10 μl): Reduced impact on processing

Determination of titer and relative aggregate content of therapeutic mAbs and other proteins that contain the Fc domain of human IgG.  It is intended primarily to monitor mAb quality during product development and production utilizing HPLC systems equipped with a fluorescence detector.

Accessories information: See the Proteometer-L Reactor, Proteometer-L Buffer Packets, and Proteometer-L Reagent series for accessory component replacement options.

Delivery information: Includes 10x Proteometer-L reagent vials, 10x Proteometer-L buffer packets, 1x reconstitution reagent. Allows for analysis up to 800 samples. This kit does NOT include a Proteometer-L Reactor.

Packaging: Supplied in a custom box with instructions for use.

Specifications

Specifications

  • Description:
    Proteometer-L consumables kit, reagent to be stored at -20 °C

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.