Order Entry
Puerto Rico
ContactUsLinkComponent
 
Virus Recombinant NiV G (from Insect)
New Product
 
undefined
Virus Recombinant NiV G (from Insect)
New Product

 

  • Protein/Peptide Type:
    Recombinant
  • Source:
    Insect
  • Species:
    Virus
  • Tag sequence:
    His Tag
  • Storage Conditions:
    −80 °C
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    Y
  • Protein Synonyms:
    Nipah Virus (NiV) Glycoprotein
  • Protein/Peptide Name:
    NiV G
  • Molecular Weight:
    ~59.5 kDa
  • Concentration:
    0.2 mg/ml
  • Formulation:
    Frozen liquid. In PBS pH 7.5
  • Nuclease-free:
    N
  • Tested Applications:
    SDS-PAGE, ELISA