ContactUsLinkComponent
VWR® Premium Series Upright Ultra-Low Temperature Freezers
VWR® Premium Series Upright Ultra-Low Temperature Freezers
Supplier:  VWR International
New Product
Certificates
undefined
VWR® Premium Series Upright Ultra-Low Temperature Freezers
Supplier:  VWR International
New Product
Specifications
  • Catalog No:
  • 77618-792
  • 77618-802
  • 77618-768
  • 77618-790
  • 77618-766
  • 77618-788
  • 77618-778
  • 77618-800
  • 77618-776
  • 77618-798
  • 77618-804
  • 77618-774
  • 77618-764
  • 77618-786
  • 77618-762
  • 77618-780
  • Doors:
  • 1 outer, 2 inner doors
  • 1 outer, 2 inner doors
  • 1 outer, 2 inner doors
  • 1 outer, 2 inner doors
  • 1 outer, 2 inner doors
  • 1 outer, 2 inner doors
  • 1 outer, 2 inner doors
  • 1 outer, 2 inner doors
  • 1 outer, 2 inner doors
  • 1 outer, 2 inner doors
  • 1 outer, 2 inner doors
  • 1 outer, 2 inner doors
  • 1 outer, 2 inner doors
  • 1 outer, 2 inner doors
  • 1 outer, 2 inner doors
  • 1 outer, 2 inner doors
  • Model:
  • Premium ULT freezer 600
  • Premium ULT freezer 700
  • Premium ULT freezer 400
  • Premium ULT freezer 600
  • Premium ULT freezer 400
  • Premium ULT freezer 600
  • Premium ULT freezer 500
  • Premium ULT freezer 700
  • Premium ULT freezer 500
  • Premium ULT freezer 700
  • Premium ULT freezer 700
  • Premium ULT freezer 500
  • Premium ULT freezer 400
  • Premium ULT freezer 600
  • Premium ULT freezer 400
  • Premium ULT freezer 500
  • Electrical:
  • 100-230 V, 50/60 Hz, NEMA 6-15P
  • 100-230 V, 50/60 Hz, NEMA 6-15P
  • 100-230 V, 50/60 Hz, NEMA 6-15P
  • 100-230 V, 50/60 Hz, NEMA 6-15P
  • 100-230 V, 50/60 Hz, NEMA 6-15P
  • 100-230 V, 50/60 Hz, NEMA 5-15P
  • 100-230 V, 50/60 Hz, NEMA 6-15P
  • 100-230 V, 50/60 Hz, NEMA 5-15P
  • 100-230 V, 50/60 Hz, NEMA 5-15P
  • 100-230 V, 50/60 Hz, NEMA 5-15P
  • 100-230 V, 50/60 Hz, NEMA 6-15P
  • 100-230 V, 50/60 Hz, NEMA 5-15P
  • 100-230 V, 50/60 Hz, NEMA 5-15P
  • 100-230 V, 50/60 Hz, NEMA 5-15P
  • 100-230 V, 50/60 Hz, NEMA 5-15P
  • 100-230 V, 50/60 Hz, NEMA 6-15P
  • Shelves/drawers:
  • 4 shelves (3 adjustable)
  • 4 shelves (3 adjustable)
  • 4 shelves (3 adjustable)
  • 4 shelves (3 adjustable)
  • 4 shelves (3 adjustable)
  • 4 shelves (3 adjustable)
  • 4 shelves (3 adjustable)
  • 4 shelves (3 adjustable)
  • 4 shelves (3 adjustable)
  • 4 shelves (3 adjustable)
  • 4 shelves (3 adjustable)
  • 4 shelves (3 adjustable)
  • 4 shelves (3 adjustable)
  • 4 shelves (3 adjustable)
  • 4 shelves (3 adjustable)
  • 4 shelves (3 adjustable)
  • Door Lock:
  • Key lock with padlock hasps
  • Key lock with padlock hasps
  • Key lock with padlock hasps
  • Key lock with padlock hasps
  • Key lock with padlock hasps
  • Key lock with padlock hasps
  • Key lock with padlock hasps
  • Key lock with padlock hasps
  • Key lock with padlock hasps
  • Key lock with padlock hasps
  • Key lock with padlock hasps
  • Key lock with padlock hasps
  • Key lock with padlock hasps
  • Key lock with padlock hasps
  • Key lock with padlock hasps
  • Key lock with padlock hasps
  • Plug:
  • NEMA 6-15P
  • NEMA 6-15P
  • NEMA 6-15P
  • NEMA 6-15P
  • NEMA 6-15P
  • NEMA 5-15P
  • NEMA 6-15P
  • NEMA 5-15P
  • NEMA 5-15P
  • NEMA 5-15P
  • NEMA 6-15P
  • NEMA 5-15P
  • NEMA 5-15P
  • NEMA 5-15P
  • NEMA 5-15P
  • NEMA 6-15P
Specifications
Chart Recorders
Product Information