Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Human Recombinant TULP1 (from E. coli)

Human Recombinant TULP1 (from E. coli)

100 µg
Catalog # ABCA_AB131704-100U
New Product
Supplier:   Abcam
Supplier #:  AB131704-100UG
Human Recombinant TULP1 (from E. coli)
Catalog # ABCA_AB131704-100U
Supplier:  Abcam
New Product
CAS Number:  
Human Recombinant TULP1 (from E. coli)
Human Recombinant TULP1 (from E. coli)

100 µg

CAS Number:  
Price
$475.00
Log in for availability
Each (100µG)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Recombinant Human TULP1 protein is a Human Fragment protein, in the 290 to 542 aa range, expressed in Escherichia coli, with >90% purity and suitable for SDS-PAGE, MS.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifications

Specifications

  • Size:
    100 µg
  • Storage Conditions:
    Store at −20 °C
  • Form:
    Liquid
  • Animal-Free:
    N
  • Purity:
    >90%
  • Tested Applications:
    MS, SDS-PAGE
  • Concentration:
    1 mg/ml
  • Source:
    E. coli
  • Cat. No.:
    ABCA_AB131704-100U
  • Shipping Temperature:
    20
  • Biological Activity:
    Standard
  • Protein/Peptide Type:
    Recombinant
  • Formulation:
    pH: 8, Constituents: 50% Glycerol (glycerin, glycerine), 1.17% Sodium chloride, 0.32% Tris HCl, 0.08% (R*,R*)-1,4-Dimercaptobutan-2,3-diol, 0.06% EDTA
  • Protein Synonyms:
    TUBL1|TULP1|Tubby-related protein 1|Tubby-like protein 1
  • Protein/Peptide Name:
    TULP1
  • Conjugation:
    Unconjugated
  • Species:
    Human

Specifications