Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Uranium Standards, 100 µg/ml

New Product
Uranium Standards, 100 µg/ml
New Product
 
Uranium Standards, 100 µg/ml
Uranium Standards, 100 µg/ml

 

 
   
 

 : ISO 17025/17034.

Product Family Options

 

  • Element
    Concentration
    Size
    Matrix
    Pack type
     
     
  • Uranium
    100 µg/ml
    125 ml
    1% HNO₃
    LDPE bottle
       
       
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $57.00
    $57.00
    Each(1/Itm/Bx)
       $57.00

     : MSU-100PPM-125ML
     
     
    New Product
    New Product
  • Uranium
    100 µg/ml
    500 ml
    1% HNO₃
    LDPE bottle
       
       
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $136.00
    $136.00
    Each(1/Itm/Bx)
       $136.00

     : MSU-100PPM-500ML
     
     
    New Product
    New Product

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.