Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
EPA Method 200.8 Internal Standards

EPA Method 200.8 Internal Standards

New Product
Supplier:   Inorganic Ventures
EPA Method 200.8 Internal Standards
Supplier:  Inorganic Ventures
New Product
Certificates
EPA Method 200.8 Internal Standards
EPA Method 200.8 Internal Standards

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Certifications: ISO 17025/17034.

Product Family Options

Product Information

  • Size
    Elements and concentration
    Matrix
    Pack type
    Availability
    Price
  • 125 ml
    20 µg/ml each of Bismuth, Scandium, Yttrium, Indium, and Terbium
    5% HNO₃
    LDPE bottle
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $134.00
    $134.00
    Each(1/Itm/Bx)
       $134.00

    Supplier Number: 2008ISS-125ML
    Specifications:
    New Product
    New Product
  • 500 ml
    20 µg/ml each of Bismuth, Scandium, Yttrium, Indium, and Terbium
    5% HNO₃
    LDPE bottle
    Log in for availability
    Log in for availability
    Each(500ml)
    Each(500ml) - $296.00
    $296.00
    Each(500ml)
       $296.00

    Supplier Number: 2008ISS-500ML
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    New Product
    New Product

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.