Anti-CD235ab Human Monoclonal Antibody [clone: HIR2] (Spark PLUS UV395™)
Supplier: Biolegend
New Product
Certificates
About this item
Spark PLUS UV395™ anti-human CD235ab [HIR2]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: FC
Each lot of this is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is ≤ 0.03 µg per million cells in 100 µL volume. It is recommended that the reagent be titrated for optimal performance for each application.; * Spark PLUS UV395™ has a maximum excitation of 355 nm and a maximum emission of 385 nm.;
Antibody type: Primary
Antigen symbol: CD235ab
Clonality: Monoclonal
Clone: HIR2
Conjugation: Spark PLUS UV395™
Epitope:
Host: Human
Isotype: IgG2b kappa
Reactivity: Human
Specifications
- Catalog No:
- 78027-020
- 78027-021
- Antigen Symbol:
- CD235ab
- CD235ab
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Spark PLUS UV395™
- Spark PLUS UV395™
- Clone:
- HIR2
- HIR2
- Host:
- Human
- Human
- Isotype:
- IgG2b kappa
- IgG2b kappa
- Flow Cytometry:
- Yes
- Yes