Anti-CD19 Rat Monoclonal Antibody [clone: 6D5] (Spark PLUS UV395™)
New Product
Spark PLUS UV395™ anti-mouse CD19 [6D5]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC
Each lot of this is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is ≤ 0.25 µg per million cells in 100 µL volume. It is recommended that the reagent be titrated for optimal performance for each application.; * Spark PLUS UV395™ has a maximum excitation of 355 nm and a maximum emission of 385 nm.;
Antibody type: Primary Antigen symbol: CD19 Clonality: Monoclonal Clone: 6D5 Conjugation: Spark PLUS UV395™ Epitope: Host: Rat Isotype: IgG2a kappa Reactivity: Mouse
- Catalog No:
- 78026-771
- 78026-772
- Antigen Symbol:
- CD19
- CD19
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- 6D5
- 6D5
- Host:
- Rat
- Rat
- Gene ID:
- 12478
- 12478
- Isotype:
- IgG2a kappa
- IgG2a kappa
- Flow Cytometry:
- Yes
- Yes
- Concentration:
- 0.20 mg/ml
- 0.20 mg/ml
- Storage Temperature:
- 2 -8 °C
- 2 -8 °C
- Shipping Temperature:
- Ambient RT
- Ambient RT